Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric acid, Environmental Grade, 67-70%, Thermo Scientific Chemicals

Catalog No. AA38747K4
Change view
Click to view available options
Quantity:
500 mL
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38747K4 2.5 L
AA38747AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA38747K4 Supplier Thermo Scientific Chemicals Supplier No. 038747K4

missing translation for 'validMainframeMsg'

Main commercial usage is in the production of fertilizers and reagent for nitration. It implies in the applications for the production of explosives, nylon precursors, and specialty organic compounds. Nitric acid indirectly used as an oxidizer in liquid-fueled rockets for determining trace level metals in solutions. It is used as a chemical doping agent for organic semiconductors, and in purification processes for raw carbon nanotubes, artificially age pine and maple.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Appearance Colorless to pale yellow liquid
Color Colorless to Yellow
Boiling Point 121°C
Density 1.41 g/cm3 at 20°C
Odor Pungent
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Assay Percent Range 67-70%
Quantity 2.5 L
Physical Form Liquid,single sub-boiling quartz distillation
UN Number 2031
Solubility Information Miscible in water and ether.
Formula Weight 63.01
Grade Environmental
Concentration or Composition (by Analyte or Components) Nitric Acid: 69.0%; Water: 31.0%
Chemical Name or Material Nitric Acid
Show More Show Less
Hazard Category H272-H290-H314-H331
Precautionary Statement P210-P220-P221-P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 231-714-2
RTECSNumber QU5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.