Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, Fuming ACS, 90%, Spectrum™ Chemical
SDP

Catalog No. 18612007 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
400 mL
2.5 L
Packaging:
Amber Glass Bottle
Poly-Coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-007 2.5 L Poly-Coated Glass Bottle
18-612-006 400 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-612-007 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. N10802.5LTPV

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7697-37-2

Specifications

CAS Max % 1
Heavy Metals 0.2 ppm
Packaging Poly-Coated Glass Bottle
Residue after Ignition 2e-05
Quantity 2.5 L
Percent Purity 90%
Grade Reagent
Chemical Name or Material Nitric Acid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.