Learn More
Specifications
Specifications
| pH | 1.0 |
| Color | Colorless Yellow |
| Melting Point | -41°C |
| Boiling Point | 120.5°C |
| Density | 1.41g/cm³ |
| Heavy Metals (as Pb) | 5ppm max. |
| Identification | Pass Test |
| Packaging | Glass Bottle |
| Vapor Pressure | 0.37 - 0.40 kPa |
| Assay Percent Range | ≥90.0% |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Oxidizer: Contact with combustible/organic material may cause fire. May cause pulmonary edema. Hygroscopic. Containers should be vented periodically in order to overcome pressure buildup. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen. NFPA Health:4 Flammability:0 Instability:0 |
| DOTInformation | DOT Class 8, : Corrosive |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.