Learn More
Sigma Aldrich Fine Chemicals Biosciences Nitric acid puriss. p.a. r1L
Catalog No. 502897397
Encompass
Nitric acid (HNO3) is a highly corrosive strong mineral acid. 10%(v/v) nitric acid SOLUTIONon is useful for cleaning laboratory glasswares. Reaction of HNO3 with various original and synthetic mineral dust/mineral oxide surfaces was studied in a low-pressure Knudsen reactor operating at 298K.
Catalog No. 50-289-7397 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 307091L
Nitric acid (HNO3) is a highly corrosive strong mineral acid. 10%(v/v) nitric acid SOLUTIONon is useful for cleaning laboratory glasswares. Reaction of HNO3 with various original and synthetic mineral dust/mineral oxide surfaces was studied in a low-pressure Knudsen reactor operating at 298K.