Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Nitric Acid Solution, 15.8M, Concentrated
SCI_ED

Catalog No. S25449A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
15.8M -70%, ACS GRADE
6M
Concentrated
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25449A 500 mL 6M
S25906 500 mL Concentrated
S25449C 2.5 L 15.8M -70%, ACS GRADE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S25449A
Add to Cart
Add to Cart

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Density 1.1 to 1.4g/mL
Packaging 6M
Quantity 500 mL
Physical Form Liquid
Linear Formula HNO3
Formula Weight 63.02
Chemical Name or Material Nitric Acid Solution, 15.8M, Concentrated
Safety handling Check Alert storage code Red
ShelfLife 2 years
DOTInformation DOT Class 8, II, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.