Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, Technical 42 Degrees Baume’, Reagents

Catalog No. 01336825
Change view
Click to view available options
Quantity:
2.2 L
600 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-825 600 lb.
01-336-826 2.2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-825 Supplier Reagents Holdings Llc Supplier No. C5101700600LB
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nitric Acid, Technical 42 Degrees Baume’, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym Azotic Acid, Hydrogen nitrate, Aqua fortis
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

pH 0
Color Colorless to Slight Yellow
Boiling Point 121°C
Packaging Steel Drum
Specific Gravity 1.4155
Quantity 600 lb.
Physical Form Liquid
Grade Technical
Chemical Name or Material Nitric Acid, Technical 42 Degrees Baume’
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.