Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, TraceGrade, 67-70%, Spectrum™ Chemical
SDP

Catalog No. 18604844 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
2.5 L
6 x 500 mL
4 x 2.5 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-844 4 x 2.5 L Poly Bottle
18-604-843 2.5 L Poly Bottle
18-604-842 6 x 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-604-844 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. N10842.5LTPLCS4
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N1084, 7697-37-2, HNO3

Spectrum™ Chemical Nitric Acid, TraceGrade is a strong, highly corrosive mineral acid with extremely low metal content (over 50 with levels less than 1 PPB). This grade is primarily used as an analytical reagent in elemental analysis determinations using ICP and AA techniques, digestion of water samples and as a doping agent for organic semiconductors.

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

CAS Max % 70%, 30%
Packaging Poly Bottle
Quantity 4 x 2.5 L
Percent Purity 67 to 70%
Grade Trace CERT
Chemical Name or Material Nitric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.