Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric acid, TRACESELECT™ Ultra, for UltraTrace Analysis, 67-69%, Solstice

Catalog No. 60044474 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
2 L
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-474 2 L Poly Bottle
60-011-10 500 mL Poly Bottle
60-044-472 1 L Poly Bottle
60-044-473 250 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-044-474 Supplier Solstice Supplier No. 026502L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TraceSELECT™ Ultra, for ultratrace analysis, 67-69%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Boiling Point 122°C
Density 1.420 g/cm3
Packaging Poly Bottle
Vapor Pressure 8mmHg (20°C)
Assay Percent Range 65 to 71%
Quantity 2 L
Linear Formula HNO3
UN Number UN2031
Formula Weight 63.01
Percent Purity 67 to 69%
Grade Ultratrace
Chemical Name or Material Nitric acid, TRACESELECT™ Ultra, for UltraTrace Analysis, 65-71%,
Show More Show Less
Hazard Statement P210-P220-P260-P280-P305 + P351 + P338-P370 + P378
Precautionary Statement H272-H290-H314
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Nitric acid
Recommended Storage Room Temp
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.