Learn More
Biotium Nitric Oxide Detection Nitric Oxide NO and Reactive Oxygen Species ROS Nitric Oxide NO Detection Chemical Reagents 1/EA
Catalog No. 5000501588
Encompass_Preferred
DAA (1 2-Diaminoanthraquinone) reacts with nitric oxide (NO) to form a red ( subEm 580 nm) fluorescent precipitate DAA has been used to detect NO concentration in rat retinas after injury to the optic nerve ul liYellow orange solid soluble in DMSO liStore desiccated at -20C liCsub14Hsub12Nsub2Osub6S liMW 336 32 /ul
Catalog No. 50-005-01588 Supplier Biotium Supplier No. 00300
DAA (1 2-Diaminoanthraquinone) reacts with nitric oxide (NO) to form a red ( subEm 580 nm) fluorescent precipitate DAA has been used to detect NO concentration in rat retinas after injury to the optic nerve ul liYellow orange solid soluble in DMSO liStore desiccated at -20C liCsub14Hsub12Nsub2Osub6S liMW 336 32 /ul