Learn More
Apexbio Technology LLC Nitric Oxide Synthase (599-613) Blocking Peptide, Bovine Endothelial Cell 25mg
SDP
Catalog No. 5000096709
Encompass_Preferred
Nitric Oxide Synthase 599-613 Blocking Peptide Bovine Endothelial Cell is a peptide inhibitor targeting Nitric Oxide Synthase (NOS) It is designed to block NOS activity thereby modulating nitric oxide (NO) synthesis and associated cellular signaling pathways Nitric Oxide Synthase 599-613 Blocking Peptide Bovine Endothelial Cell exerts its biological activity primarily by interfering with the enzymatic function of NOS resulting in reduced NO formation Based on these pharmacological properties Nitric Oxide Synthase 599-613 Blocking Peptide Bovine Endothelial Cell holds research potential in studies of ischemia-reperfusion injury cytokine-mediated inflammation and antihypertensive mechanism evaluation
Catalog No. 50-000-96709 Supplier Apexbio Technology LLC Supplier No. A111225
Add to Cart
Add to Cart
Nitric Oxide Synthase 599-613 Blocking Peptide Bovine Endothelial Cell is a peptide inhibitor targeting Nitric Oxide Synthase (NOS) It is designed to block NOS activity thereby modulating nitric oxide (NO) synthesis and associated cellular signaling pathways Nitric Oxide Synthase 599-613 Blocking Peptide Bovine Endothelial Cell exerts its biological activity primarily by interfering with the enzymatic function of NOS resulting in reduced NO formation Based on these pharmacological properties Nitric Oxide Synthase 599-613 Blocking Peptide Bovine Endothelial Cell holds research potential in studies of ischemia-reperfusion injury cytokine-mediated inflammation and antihypertensive mechanism evaluation