Learn More
Hach Company Nitrification Control Test Kit
Supplier:  Hach Company 2882200
Encompass
The Nitrification Control Kit is used for the routine monitoring of water quality parameters that contribute to or indicate nitrification is occurring in chloraminated water systems. This portable and versatile kit includes a DR 900 Colorimeter and reagents for Free Ammonia and Monochloramine, Nitrite, and Total Chlorine tests.
Catalog No. 50-199-6766