Learn More
Hach Company Nitrification Inhibitor for BOD, Formula 2533™, TCMP, 500 g
Catalog No. 501994798 Shop All Hach Company Products
Encompass
TCMP - 2-chloro-6 (trichloromethyl) pyridine (N-Serve), coated on an inert substrate. Use 0.16 gr in a 300-mL sample to inhibit nitrogenous oxygen demand in the BOD test (Use Dispenser Cap, Part No. 45901), 500 g.
Catalog No. 50-199-4798 Supplier Hach Company Supplier No. 253334
TCMP - 2-chloro-6 (trichloromethyl) pyridine (N-Serve), coated on an inert substrate. Use 0.16 gr in a 300-mL sample to inhibit nitrogenous oxygen demand in the BOD test (Use Dispenser Cap, Part No. 45901), 500 g.