Learn More
Nitrile Examination Disposable Gloves
Catalog No. NGPF7000XL Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
NGPF7000XL X-Large
NGPF7000M Medium
NGPF7000L Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. NGPF7000XL
This item is not returnable. View return policy
Disposable nitrile gloves, powder free, non-sterile, ambidextrous
  • 3.5 gm nitrile gloves
  • Ambidextrous with beaded cuff
  • Color: blue
  • Certifications: 510K, CE Certification, ISO9001 & ISO13485
  • Impermeability as per EN455-1: AQL 1.5
Specifications
AQL 1.5
Color Blue
Disposable Single-use
Material Nitrile
Size X-Large
FDA 510 (k) Yes