Learn More
Nitrile Examination Gloves
Catalog No. 12888550C Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Large
Catalog No. Size
12888550C Large
1 options
This item is not returnable. View return policy
Catalog No. 12888550C
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Non-sterile, powder free, nitrile examination gloves.
  • Nitrile
  • Ambidextrous with beaded cuff
  • Color: Blue
  • Finger textured
  • ASTM D6319
Specifications
Certifications/Compliance ASTM D6319
Color Blue
Disposable Single-use
Finger Thickness (English) 3.1 to 5.5 mil
Finger Thickness (Metric) 0.08 to 0.14 mm
Material Nitrile
Palm Thickness (English) 2.4 to 3.5 mil
Palm Thickness (Metric) 0.06 to 0.09 mm
Size Large
Thickness (English) 3.1 to 5.5 mil
Thickness (Metric) 0.08 to 0.14 mm