Learn More
Chemglass Life Sciences Nitrile Gloves, Black, Examination, Bold, Large
SDP
Catalog No. 501531242
Encompass_Preferred
Nitrile Gloves, Black, Examination, Bold, Large. Nitrile provides better elasticity than vinyl, better puncture resistance than latex or vinyl, and better abrasion resistance than chloroprene. The Bold nitrile, powder-free, disposable glove has better puncture resistance compared to latex or vinyl gloves. Finger-textured finish for improved gripping action. This glove has a palm thickness of 4.5 mils and a finger thickness of 5 mils, and is 9.4" long from the tip of the middle finger to the edge of the beaded wrist cuff. Nitrile provides better elasticity than vinyl, better puncture resistance than latex or vinyl, better abrasion resistance than chloroprene, and reduces the likelihood of allergic reaction. This powder-free glove leaves no residue, reducing the risk of contamination compared to a powdered glove. This ambidextrous black glove is suitable for many applications.
This item is not returnable. View return policy
Catalog No. 50-153-1242 Supplier Chemglass Life Sciences Supplier No. CG1790B04
Nitrile Gloves, Black, Examination, Bold, Large. Nitrile provides better elasticity than vinyl, better puncture resistance than latex or vinyl, and better abrasion resistance than chloroprene. The Bold nitrile, powder-free, disposable glove has better puncture resistance compared to latex or vinyl gloves. Finger-textured finish for improved gripping action. This glove has a palm thickness of 4.5 mils and a finger thickness of 5 mils, and is 9.4" long from the tip of the middle finger to the edge of the beaded wrist cuff. Nitrile provides better elasticity than vinyl, better puncture resistance than latex or vinyl, better abrasion resistance than chloroprene, and reduces the likelihood of allergic reaction. This powder-free glove leaves no residue, reducing the risk of contamination compared to a powdered glove. This ambidextrous black glove is suitable for many applications.