Learn More
Chemglass Life Sciences Nitrile Gloves, Blue, Examination, Robust, Large
SDP
Catalog No. 501531246
Encompass_Preferred
Nitrile Gloves, Blue, Examination, Robust, Large. Nitrile provides better elasticity than vinyl, better puncture resistance than latex or vinyl, and better abrasion resistance than chloroprene. Powder-free, ambidextrous, disposable gloves with better puncture resistance than latex or vinyl. Textured along the full length of the fingers for improved grip. Palm thickness is 5 mils and finger thickness is 6 mils. Gloves measure 9.4" long from tip of middle finger to edge of beaded wrist cuff which is 2 mils thick. Nitrile has better elasticity than vinyl, better puncture resistance than latex and vinyl, and better abrasion resistance than chloroprene. Reduced likelihood of allergic reaction compared to latex. Powder-free means no residue and reduced risk of contamination.
This item is not returnable. View return policy
Catalog No. 50-153-1246 Supplier Chemglass Life Sciences Supplier No. CG1790R04
Nitrile Gloves, Blue, Examination, Robust, Large. Nitrile provides better elasticity than vinyl, better puncture resistance than latex or vinyl, and better abrasion resistance than chloroprene. Powder-free, ambidextrous, disposable gloves with better puncture resistance than latex or vinyl. Textured along the full length of the fingers for improved grip. Palm thickness is 5 mils and finger thickness is 6 mils. Gloves measure 9.4" long from tip of middle finger to edge of beaded wrist cuff which is 2 mils thick. Nitrile has better elasticity than vinyl, better puncture resistance than latex and vinyl, and better abrasion resistance than chloroprene. Reduced likelihood of allergic reaction compared to latex. Powder-free means no residue and reduced risk of contamination.