Learn More
Mascon Medical Mascon Medical™ Nitrile Gloves Case, Latex Free, Powder Free, Textured Fingers, ASTM D6319, Size Small, 300EA/Box, 1 Case/10 Boxes, Single Use
SDP
Catalog No. NC2540611
Encompass_Preferred
Mascon Medical™ Nitrile Gloves Case are perfect for health care settings, first aid, pet care and diabetic care. These excellent latex-free, powder-free barrier protection gloves have textured fingers, are flexible, comfortable and soft to the touch. They are available in a range of sizes from size extra small through to size extra large and meet ASTM standard D6319. Specifications:• Size: Small• Qty. / Case: 3000• Material: Nitrile• Color: Blue• Free From: Latex, Powder• ASTM Standard: D6319• Packaging: 300 EA/Box, 10 Boxes/Case
Catalog No. NC2540611 Supplier Mascon Medical Supplier No. MMGLN300SCS
Mascon Medical™ Nitrile Gloves Case are perfect for health care settings, first aid, pet care and diabetic care. These excellent latex-free, powder-free barrier protection gloves have textured fingers, are flexible, comfortable and soft to the touch. They are available in a range of sizes from size extra small through to size extra large and meet ASTM standard D6319. Specifications:• Size: Small• Qty. / Case: 3000• Material: Nitrile• Color: Blue• Free From: Latex, Powder• ASTM Standard: D6319• Packaging: 300 EA/Box, 10 Boxes/Case