Learn More
MSC NITRILE GLOVES LINED 13
Catalog No. 5000793579
Encompass
MCR Safety Nitrile Chemical-Resistant Gloves are engineered to provide robust protection in various industrial applications. They offer superior resistance to many chemicals and are three times more puncture-resistant than traditional rubber gloves, making them an excellent choice for individuals with latex allergies.
  • Made from durable nitrile material
  • Features a 15 mil thickness for enhanced protection
  • Flock lined for comfort and easy donning
  • Provides 13 inch length for extended wrist and forearm coverage
  • Diamond grip ensures secure handling of objects
  • Straight cuff design for easy wear
  • Component materials comply with all federal regulations for food contact
  • FDA compliant, suitable for various regulated environments
Catalog No. 50-007-93579 Supplier MSC Supplier No. 84027283
MCR Safety Nitrile Chemical-Resistant Gloves are engineered to provide robust protection in various industrial applications. They offer superior resistance to many chemicals and are three times more puncture-resistant than traditional rubber gloves, making them an excellent choice for individuals with latex allergies.
  • Made from durable nitrile material
  • Features a 15 mil thickness for enhanced protection
  • Flock lined for comfort and easy donning
  • Provides 13 inch length for extended wrist and forearm coverage
  • Diamond grip ensures secure handling of objects
  • Straight cuff design for easy wear
  • Component materials comply with all federal regulations for food contact
  • FDA compliant, suitable for various regulated environments