Learn More
Chemglass Life Sciences Nitrile Gloves, Medium, Extra Thick (6 mil), Examination
SDP
Catalog No. 5000980691
Encompass_Preferred
6. 5gm Powder-Free Nitrile Disposable Glove. Ambidextrous. Non-Sterile. 240mm Length. Fingertip Textured. ASTM Standard.
Catalog No. 50-009-80691 Supplier Chemglass Life Sciences Supplier No. CG179103
6. 5gm Powder-Free Nitrile Disposable Glove. Ambidextrous. Non-Sterile. 240mm Length. Fingertip Textured. ASTM Standard.