Learn More
Chemglass Life Sciences Chemglass Nitrile Examination Gloves, Small (Pack of 100)
SDP
Catalog No. NC1863270
Encompass_Preferred
Powder-free nitrile examination gloves designed to provide tactile sensitivity, puncture resistance, and a comfortable fit for clinical and laboratory tasks. Suitable for users with latex allergies, these gloves support easy donning and help reduce roll-back.
  • Powder-free and ultra-soft nitrile material.
  • Ambidextrous design for ease of use.
  • Palm thickness: 4 mil; finger thickness: 5 mil.
  • Pack size: 100 gloves per pack.
  • Intended for examination use; meets 21 CFR 880.6250 Class 1 requirements.
  • Small size: palm width 80±10 mm; length ≥220 mm.
  • Typical weight: 2.76-3.01 g per glove.
  • Good heat, puncture, and abrasion resistance.
This item is not returnable. View return policy
Catalog No. NC1863270 Supplier Chemglass Life Sciences Supplier No. CG179002
Powder-free nitrile examination gloves designed to provide tactile sensitivity, puncture resistance, and a comfortable fit for clinical and laboratory tasks. Suitable for users with latex allergies, these gloves support easy donning and help reduce roll-back.
  • Powder-free and ultra-soft nitrile material.
  • Ambidextrous design for ease of use.
  • Palm thickness: 4 mil; finger thickness: 5 mil.
  • Pack size: 100 gloves per pack.
  • Intended for examination use; meets 21 CFR 880.6250 Class 1 requirements.
  • Small size: palm width 80±10 mm; length ≥220 mm.
  • Typical weight: 2.76-3.01 g per glove.
  • Good heat, puncture, and abrasion resistance.