Learn More
Chemglass Life Sciences Nitrile Gloves, Trans-Blue, Examination, Transform, Small
SDP
Catalog No. 5000980653
Encompass_Preferred
Features: Material: Nitrile (100% Latex-Free). Type: Non-Sterile. Glove Design and Feature: Ambidextrous and Finger Textured. Cuff: Beaded. Usage: For Single Use Only. Color: Trans-Blue. ASTM 6978. Chemo Rated. Thickness: 3. 2 mil palm and 4mil fingertip thickness. Packaging:  200 pieces per box. Automated packing to enhance single glove dispensing
Catalog No. 50-009-80653 Supplier Chemglass Life Sciences Supplier No. CG1790T02
Features: Material: Nitrile (100% Latex-Free). Type: Non-Sterile. Glove Design and Feature: Ambidextrous and Finger Textured. Cuff: Beaded. Usage: For Single Use Only. Color: Trans-Blue. ASTM 6978. Chemo Rated. Thickness: 3. 2 mil palm and 4mil fingertip thickness. Packaging:  200 pieces per box. Automated packing to enhance single glove dispensing