Learn More
Bellco Glass, Inc. Nitrile gloves with oats extractions, powder free, a patented coating recognized by the FDA as a skin protectant, M, 100/pk, 1000/cs
SDP
Catalog No. 502329253
Encompass_Preferred
Nitrile gloves with oats extractions, powder free, a patented coating recognized by the FDA as a skin protectant, M, 100/pk, 1000/cs. Nitrile gloves are a type of disposable glove made from synthetic rubber, this means there is no risk of latex allergies. They are the most popular glove type in our range and offer superior strength, dexterity and resistance to oils and aqueous chemicals in comparison to vinyl or latex. For these reasons, they are often used in the medical, laboratory and manufacturing industries. Oats extractions is a patented coating recognized by the FDA as a skin protectan. Clinical studies have shown that colloidal oatmeal could reduce and control rash, dry skin. • Single use only• Made in Malaysia• Ambidextrous• Beaded cuff• Finger-textured• Powder free• Chlorinated• Tensile Strength: Minimum 18.0 MPa( Before Aging)• Palm thickness: 0.05mm
Catalog No. 50-232-9253 Supplier Bellco Glass, Inc. Supplier No. 451400657
Nitrile gloves with oats extractions, powder free, a patented coating recognized by the FDA as a skin protectant, M, 100/pk, 1000/cs. Nitrile gloves are a type of disposable glove made from synthetic rubber, this means there is no risk of latex allergies. They are the most popular glove type in our range and offer superior strength, dexterity and resistance to oils and aqueous chemicals in comparison to vinyl or latex. For these reasons, they are often used in the medical, laboratory and manufacturing industries. Oats extractions is a patented coating recognized by the FDA as a skin protectan. Clinical studies have shown that colloidal oatmeal could reduce and control rash, dry skin. • Single use only• Made in Malaysia• Ambidextrous• Beaded cuff• Finger-textured• Powder free• Chlorinated• Tensile Strength: Minimum 18.0 MPa( Before Aging)• Palm thickness: 0.05mm