Learn More
MSC Milwaukee Tool Cut-Resistant Dipped Gloves, 18 ga, 2XL, 10 in., 1 ea.
Catalog No. 5000687678
Encompass
These cut-resistant gloves are designed for high dexterity, making them ideal for handling small objects in various work environments. The 18-gauge knit is constructed from high-performance polyethylene to provide reliable cut protection while maintaining a breathable, comfortable fit for all-day use. They feature a durable, long-lasting nitrile dip on the palm and fingers, along with a reinforced nitrile coating between the thumb and index finger for added strength in high-wear areas.
  • ANSI/ISEA 105-2016 cut level A2 protection
  • Touchscreen compatible fingertips for operating devices without removing gloves
  • Seamless knit design with high dexterity for precision tasks
  • Breathable materials ensure comfort during extended wear
  • Color-coded band for quick and easy cut level identification
Catalog No. 50-006-87678 Supplier MSC Supplier No. 65431991
These cut-resistant gloves are designed for high dexterity, making them ideal for handling small objects in various work environments. The 18-gauge knit is constructed from high-performance polyethylene to provide reliable cut protection while maintaining a breathable, comfortable fit for all-day use. They feature a durable, long-lasting nitrile dip on the palm and fingers, along with a reinforced nitrile coating between the thumb and index finger for added strength in high-wear areas.
  • ANSI/ISEA 105-2016 cut level A2 protection
  • Touchscreen compatible fingertips for operating devices without removing gloves
  • Seamless knit design with high dexterity for precision tasks
  • Breathable materials ensure comfort during extended wear
  • Color-coded band for quick and easy cut level identification