Learn More
Nitrile Powder Free Gloves
Catalog No. 10321110 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
10-321-110 Large
10-319-110 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-321-110
This item is not returnable. View return policy
Textured nitrile multi-use gloves
  • Violet blue
  • Powder free
  • Tested for use with chemotherapy drugs
  • Textured
  • Thickness (finger and palm): 0.05 mm, 1.96 mils
Specifications
Color Blue
Disposable Single-use
Material Nitrile
Powdered Powder Free
Size Large
Thickness (English) 2 mil
Thickness (Metric) 0.05 mm