Learn More
MSC GE Lighting Multi-Purpose Work Gloves, Nitrile Coated, Cut Resistant, XL, 1/Pr.
Catalog No. 5000687832
Encompass
These multi-purpose work gloves feature a 13-gauge liner made of HPPE, polyester, glass fiber, steel wire, and spandex to provide robust hand protection. Designed for versatile use in general work and electronics industries, they incorporate TPR impact protection and a micro-foam nitrile coating for superior grip and abrasion resistance.
  • ANSI/ISEA cut level A4 rating for high protection
  • Micro-foam nitrile coating offers chemical and oil resistance
  • Reinforced thumb crotch for added durability
  • Longer cuff design for enhanced protection and comfort
  • Touchscreen compatible for ease of use
  • CE certified and compliant with EN388-4X42D standards
Catalog No. 50-006-87832 Supplier MSC Supplier No. 33252438
Add to Cart
Add to Cart
These multi-purpose work gloves feature a 13-gauge liner made of HPPE, polyester, glass fiber, steel wire, and spandex to provide robust hand protection. Designed for versatile use in general work and electronics industries, they incorporate TPR impact protection and a micro-foam nitrile coating for superior grip and abrasion resistance.
  • ANSI/ISEA cut level A4 rating for high protection
  • Micro-foam nitrile coating offers chemical and oil resistance
  • Reinforced thumb crotch for added durability
  • Longer cuff design for enhanced protection and comfort
  • Touchscreen compatible for ease of use
  • CE certified and compliant with EN388-4X42D standards