Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrilotriacetic Acid (Certified ACS), Fisher BioReagents

Catalog No. BP2670100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP2670100 100 g Poly Bottle
BP2670500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BP2670100
This item is not returnable. View return policy
Nitrilotriacetic Acid is a chelating and sequestering agent. A builder in synthetic detergents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-13-9
Molecular Formula C6H9NO6
Molecular Weight (g/mol) 191.14
MDL Number MFCD00004287
InChI Key MGFYIUFZLHCRTH-UHFFFAOYSA-N
Synonym nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i
PubChem CID 8758
ChEBI CHEBI:44557
IUPAC Name 2-[bis(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CC(O)=O)CC(O)=O

Specifications

Melting Point 245°C
Color White
pH 2.3
Assay Percent Range ≥98.0 %
Packaging Poly Bottle
Linear Formula N(CH2COOH)3
Quantity 100 g
Merck Index 15, 6665
Formula Weight 191.14
Percent Purity ≥98.0%
Grade Certified ACS
Physical Form Powder or Solid
Chemical Name or Material Nitrilotriacetic Acid
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Signal Word: WARNING!
Precautionary Statement Emergency Overview
Harmful if swallowed. Irritating to eyes and skin. Suspect cancer hazard.
NFPA
Health: 2
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.