Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrilotriacetic Acid Disodium Salt 99.0+%, TCI America™
SDP

Catalog No. N0100500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0100500G 500 g
N010025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0100500G Supplier TCI America Supplier No. N0100500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15467-20-6
Molecular Formula C6H7NNa2O6
Molecular Weight (g/mol) 235.103
MDL Number MFCD00064230
InChI Key RHPXYIKALIRNFA-UHFFFAOYSA-L
Synonym nitrilotriacetic acid disodium salt, kiresuto ntb, disodium nitrilotriacetate, nitrilotriacetic acid, disodium salt, disodium hydrogen nitrilotriacetate, unii-llv4ki749r, disodium nitriloacetate, nitrilotriacetic acid,disodium salt, llv4ki749r, n,n-bis carboxymethyl glycine disodium salt
PubChem CID 27301
IUPAC Name disodium;2-[carboxylatomethyl(carboxymethyl)amino]acetate
SMILES C(C(=O)O)N(CC(=O)[O-])CC(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 500 g
Formula Weight 235.10
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nitrilotriacetic Acid Disodium Salt
EINECSNumber (2)-1277
RTECSNumber AJ0350000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.