Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrilotriacetic acid, Solstice

Catalog No. 6002519 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
50 g
25 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-025-19 25 kg
60-025-20 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-025-19 Supplier Solstice Supplier No. 3453925KG
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

for complexometry,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-13-9
Molecular Formula C6H9NO6
Molecular Weight (g/mol) 191.14
MDL Number MFCD00004287
InChI Key MGFYIUFZLHCRTH-UHFFFAOYSA-N
Synonym nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i
PubChem CID 8758
ChEBI CHEBI:44557
IUPAC Name 2-[bis(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CC(O)=O)CC(O)=O

Specifications

Flash Point 100°C (212°F)
Linear Formula N(CH2COOH)3
Quantity 25 kg
UN Number NONH for all modes of transport
Beilstein 1710776
Formula Weight 191.14g/mol
Percent Purity ≥99%
Chemical Name or Material Nitrilotriacetic acid
Hazard Statement P281-P305 + P351 + P338
RTECSNumber AJ0175000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.