Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Nitrilotriacetic acid trisodium salt monohydrate

Catalog No. 501766268
Encompass
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-176-6268 1 kg
50-176-6269 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-176-6268 Supplier Sigma Aldrich Supplier No. 725651KG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Nitrilotriacetic acid trisodium salt monohydrate
TRUSTED_SUSTAINABILITY

Specifications

CAS 18662-53-8
Melting Point >300°C (lit.)
Molecular Formula C6H6NNa3O6 -+ H2O
Linear Formula N(CH2COONa)3 -+ H2O
MDL Number MFCD00150789
Quantity 1 kg
Synonym Sodium nitrilotriacetate tribasic monohydrate
Molecular Weight (g/mol) 275.1
Percent Purity ≥98.0% (KT)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.