Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Nitrilotris(methylene)triphosphonic acid solution

Catalog No. 1447971KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-479-71KG 1 kg
14-479-7100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-479-71KG Supplier Sigma Aldrich Supplier No. 1447971KG
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Nitrilotris(methylene)triphosphonic acid solution is an antiscalant and can be removed form membrane concentrates by iron-coated waste filtration sand
  • It is a cement retarder
  • It reacts with the aluminum surface to form a coating which is an effective inhibitor of the reaction of evaporated aluminum thin films on glass or silicon with deionized water.nitrilotris(methylene)triphosphonic acid solution was used to study the mechanism of inhibition of cement hydration by phosphonic acid

Specifications

Density 1.3 g/mL (at 25°C)
Molecular Formula C3H12NO9P3
Linear Formula N[CH2P(O)(OH)2]3
MDL Number MFCD00002138
Quantity 1 kg
Synonym Nitrilotri(methylphosphonic acid) solution
Molecular Weight (g/mol) 299.05
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.