Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety NitriShield™ with Grippaz™ Technology, Orange Nitrile Gloves

Catalog No. 19812746 Shop All MCR Safety Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-812-746 Medium
19-812-745 Large
19-812-748 X-Large
19-812-749 2X-Large
19-812-747 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-812-746 Supplier MCR Safety Supplier No. 6016OM
Add to Cart
Add to Cart
This item is not returnable. View return policy

The unique fish-scale pattern of these orange nitrile gloves provides unsurpassed grip and durability. Ergonomically designed for wet grip on the outside and to reduce hand slippage on the inside.

  • 6 mil, Orange Powder Free Nitrile
  • Fish-scale pattern grip
  • Textured inside for better traction
  • 9.5 Inch Industrial/food service grade
  • Component materials comply with all relevant federal regulations for food contact

Specifications

Certifications/Compliance FDA Compliant
Applications Industrial
AQL 1.5
Color Orange
Disposable Single-use
Exterior Finish Fully Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Medium
Sterility Non-sterile
Thickness (English) 6 mil
Thickness (Metric) 0.15 mm
Model 6016OM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.