Learn More
Nitrite, Quant™ Test Strips
Catalog No. AA23283QA
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
AA23283QA 1 Kit
1 options
Catalog No. AA23283QA Supplier Thermo Scientific Chemicals Supplier No. 023283QA
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Range Measures 1 to 50ppm NO2- in aqueous solutio
Type Nitrite test strip