Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Nitrite Reducing Bacteria (NRB) Tube Test

Catalog No. 15398344 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-398-344 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-398-344 Supplier Lovibond™ Supplier No. 56B010910
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Nitrite Reducing Bacteria (NRB) Tube Test is used to monitor the levels of NRB in water systems which use nitrite as a corrosion inhibitor. The test contains a pale orange medium which reacts to metabolism by NRB’s to produce a semi-quantitative result within 5 days.

  • The Nitrite Reducing Bacteria (NRB) test is designed to assess the contamination of water samples with nitrite reducing bacteria.
  • The NRB test is used to monitor the levels of NRB in water systems with specific relevance to systems which use nitrite as a corrosion inhibitor.
  • NRB's are able to metabolize nitrite into ammonia and nitrogen gas which are aggressive corrosive agents, as well as this the corrosion inhibition of nitrite is lost.
  • Results are to be determined by the color change seen on day 1 through to 5.

Specifications

Color Pale Orange
Detects Nitrite Reducing Bacteria
Target Organism Class Bacteria
Test Type Water Monitoring
Formulation Semi-solid Pale Orange Medium for The Analysis of Anaerobic Microorganisms Capable of Nitrite Ammonification
For Use With (Application) Determine aerobic and anaerobic bacteria levels
For Use With (Equipment) DI10 Digital Incubator
Incubation Conditions 35°C
Incubation Time 5 Days
Manual/Automated Manual
No. per Pack 10/Pk.
Packaging Type Pack
Quantity 1 Ea.
Specimen Type Water, Liquids
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.