Learn More
AccuStandard, Inc NITRITE REF STD
SDP
Catalog No. NC9213398
Encompass
Anion Standard-Nitrite as Nitrogen; 500mL; Concentration: 100ug/mL in Water; CAS: NO2-N; Storage Condition: Refrig (0 deg. to 5 deg.C)
Catalog No. NC9213398 Supplier AccuStandard, Inc Supplier No. ICNO2N1X5
Anion Standard-Nitrite as Nitrogen; 500mL; Concentration: 100ug/mL in Water; CAS: NO2-N; Storage Condition: Refrig (0 deg. to 5 deg.C)