Learn More
CHEMetrics™ by AquaPhoenix Nitrite Refill Kit



Description
- The Nitrite CHEMets™ test kit employs the azo dye method.
- In an acidic solution, nitrite diazotizes with the primary aromatic amine N-(1-naphthyl)ethylenediamine dihydrochloride (NED) and then couples with sulfanilic acid to produce a highly colored azo dye. The resulting pink color is proportional to the nitrite concentration in the sample.
Specifications
Specifications
| pH Range | 7 |
| Volume (Metric) | 0.5 mL |
| For Use With (Equipment) | K-7006 |
| No. of Tests | 30 |
| Type | Nitrite Refill Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.