Learn More
ERA WATERS CORP NITRITE STANDARD 1000PPM 25ML
SDP
Catalog No. NC9733551
Encompass
One 125 mL standard containing Nitrate as N at 1000 mg/L in H2O. This CRM is covered by ERA's ISO Guide 34 accreditation.
Catalog No. NC9733551 Supplier ERA WATERS CORP Supplier No. 053
One 125 mL standard containing Nitrate as N at 1000 mg/L in H2O. This CRM is covered by ERA's ISO Guide 34 accreditation.