Learn More
ERA WATERS CORP NITRITE STD 1000MG/L 500ML
SDP
Catalog No. NC9738456
Encompass
One 500 mL standard containing Nitrite as N at 1000 mg/L in H2O. This CRM is covered by ERA's ISO Guide 34 accreditation.
Catalog No. NC9738456 Supplier ERA WATERS CORP Supplier No. 990
One 500 mL standard containing Nitrite as N at 1000 mg/L in H2O. This CRM is covered by ERA's ISO Guide 34 accreditation.