Learn More
Hach Company Hach Nitrite Standard, 100 mg/L NO₂ (30.5 mg/L NO₂-N), 500 mL (LCW965)
Catalog No. NC3058704 Shop All Hach Company Products
Encompass
Ready-to-use nitrite calibration standard prepared at a nominal concentration of 100 mg/L NO₂ (30.5 mg/L NO₂-N) in a 500 mL solution for instrument calibration and quality control.
  • Ready-to-use 500 mL solution.
  • Nominal concentration 100 mg/L NO₂ (30.5 mg/L NO₂-N).
  • Intended for instrument calibration and quality control.
  • Safety Data Sheet (SDS) available from the manufacturer.
  • Store and handle according to the SDS and local regulations.
Catalog No. NC3058704 Supplier Hach Company Supplier No. LCW965
Ready-to-use nitrite calibration standard prepared at a nominal concentration of 100 mg/L NO₂ (30.5 mg/L NO₂-N) in a 500 mL solution for instrument calibration and quality control.
  • Ready-to-use 500 mL solution.
  • Nominal concentration 100 mg/L NO₂ (30.5 mg/L NO₂-N).
  • Intended for instrument calibration and quality control.
  • Safety Data Sheet (SDS) available from the manufacturer.
  • Store and handle according to the SDS and local regulations.