Learn More
Hach Company Nitrite TNTplus Vial Test, LR (0.015-0.600 mg/L NO₂-N), 25 Tests
Catalog No. 501998132 Shop All Hach Company Products
Encompass
Nitrite TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Nitrite Nitrogen by the Diazotization method. EPA compliant.
Catalog No. 50-199-8132 Supplier Hach Company Supplier No. TNT839
Nitrite TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Nitrite Nitrogen by the Diazotization method. EPA compliant.