Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Nitrite VHR Liquid Reagent

Catalog No. 01549784 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-549-784 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-549-784 Supplier Lovibond™ Supplier No. 471170
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Nitrite VHR Liquid Reagent is designed to test and monitor high levels (up to 2500 mg/L) in a water sample.

  • Reagent designed to cover full range of nitrite dosing used in closed loop cooling water systems
  • Liquid reagent dissolves and mixes easily into sample
  • Works well in water matrix that contain glycol-based anti-freeze
  • Easily monitor nitrite levels in closed-loop cooling systems with this liquid reagent. Proper control of nitrite levels prevents pipework corrosion. This easy-to-use colorimetric method test can monitor high levels (up to 2500 mg/L) of nitrite without the need to dilute samples.
  • Unlike similar colorimetric tests on the market, this test provides the full testing range industrial water treaters need and avoids the release of potentially harmful nitrogen oxides during testing. With over 50 years of manufacturing high-quality reagents, Lovibond™ knows choosing the right test for your process or application is critical for accurate results.

Specifications

Analytes Nitrite
Format Liquid Reagent
For Use With (Application) Closed-Loop Cooling Water Systems
For Use With (Equipment) Select Lovibond™ Colorimeters and Spectrophotometers
Includes 1 x Nitrite VHR L, 500 mL
No. of Tests ∼500
No. per Pack 500 mL
Packaging Amber Glass Bottle
Quantity 1 Ea.
Range 25 to 2500 mg/L NO2-
Sample Size 10 mL
Test Method Lovibond™ M271
Test Time <5 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.