Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics™ by AquaPhoenix Nitrite Water Quality Visual Test Kit, 0 to 2000 ppm
SDP

Catalog No. 13299617
Change view
Click to view available options
No. of Tests:
30
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Tests
13-299-617 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-299-617 Supplier CHEMetrics™ by AquaPhoenix Supplier No. K7030C
Add to Cart
Add to Cart

Nitrite visual test kit featuring self-filling ampoules with pre-measured reagent for a single test. Compare results to supplied color standards to determine concentration.

  • The K-7030C Nitrite HR CHEMets™ Test Kit (0-2000 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): Refill, High Range Comparator, Acidifier Solution, 25 mL sample cup, 10 uL fixed volume pipet, pipet tips, and instructions.

Specifications

Analytes Nitrate
pH Range 7
Volume (Metric) 0.5 mL
No. of Tests 30
Range 0 to 2500 ppm
Type Water Quality Visual Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.