Learn More
Hach Company NitriVer 2 Nitrite Reagent Powder Pillows, 10 mL, pk/100
Catalog No. 501995421 Shop All Hach Company Products
Encompass
For determination of high range nitrite by the NitriVer 2 ferrous sulfate method. Hach method 8153. Range: 2 to 250 mg/L. Sample Size: 10 mL. Pack of 100 Powder Pillows.
Catalog No. 50-199-5421 Supplier Hach Company Supplier No. 2107569
For determination of high range nitrite by the NitriVer 2 ferrous sulfate method. Hach method 8153. Range: 2 to 250 mg/L. Sample Size: 10 mL. Pack of 100 Powder Pillows.