Learn More
Hach Company NitriVer 2 Nitrite Reagent Powder Pillows, pk/100
Supplier:  Hach Company 181369
Encompass
For determination of high range nitrite by the ferrous sulfate method. Sample Size: 5 mL. pk/100 Powder Pillows.
Catalog No. 50-199-4607