Learn More
Hach Company NitriVer 2 Nitrite Reagent Powder Pillows, 25 mL, pk/100
Catalog No. 501994727 Shop All Hach Company Products
Encompass
For high range nitrite determination by the ferrous sulfate method. Hach method 8153. Range::2 - 150 mg/L NO2-N. Sample Size: 25 mL. Pack of 100 powder pillows.
Catalog No. 50-199-4727 Supplier Hach Company Supplier No. 221969
For high range nitrite determination by the ferrous sulfate method. Hach method 8153. Range::2 - 150 mg/L NO2-N. Sample Size: 25 mL. Pack of 100 powder pillows.