Learn More
Hach Company NitriVer 3 Nitrite Reagent Powder PIllows, 10 mL, pk/100
Catalog No. 501995415 Shop All Hach Company Products
Encompass
For determination of low range nitrite by the NitriVer 3 diazotization method. USEPA accepted for reporting wastewater analysis using Hach Method 8507. Range: up to 0.30 mg/L NO2-N. Sample Size:10 mL. Pack of 100 Powder Pillows
Catalog No. 50-199-5415 Supplier Hach Company Supplier No. 2107169
For determination of low range nitrite by the NitriVer 3 diazotization method. USEPA accepted for reporting wastewater analysis using Hach Method 8507. Range: up to 0.30 mg/L NO2-N. Sample Size:10 mL. Pack of 100 Powder Pillows