Learn More
Hach Company NitriVer 3 Nitrite Reagent Powder Pillows, 25 mL, pk/100
Catalog No. 501994912 Shop All Hach Company Products
Encompass
For low range nitrite determination by the NitriVer 3 Diazotization Method: EPA Compliant - Hach Method 8507. Range: up to 0.300 mg/L NO₂-N Sample Size: 25 mL. Pack of 100 powder pillows.
Catalog No. 50-199-4912 Supplier Hach Company Supplier No. 1406599
For low range nitrite determination by the NitriVer 3 Diazotization Method: EPA Compliant - Hach Method 8507. Range: up to 0.300 mg/L NO₂-N Sample Size: 25 mL. Pack of 100 powder pillows.