Learn More
Hach Company NitriVer 3 Nitrite Reagent Powder Pillows, 5 mL, pk/100
Supplier:  Hach Company 1407899
Encompass
For nitrite determination by the NitriVer 3 diazotization method. Sample size 5 mL. For use with Hach visual test kits. Range: up to 0.5 mg/L NO2-N.
Catalog No. 50-199-4920