Learn More
MSC Magid Work Glove, DuraBlend Shell, NitriX Grip, A5 Cut, 1/Pr.
Catalog No. 5000688894
Encompass
These size 7, 15-gauge seamless machine knit gloves feature a high-visibility yellow DuraBlend™ shell for breathability and comfort. The sandy NitriX® Grip palm coating ensures a secure grip in wet, dry, or oily applications, while the padded palm provides impact dampening for improved control. Designed for use in assembly, manufacturing, and material handling, these gloves provide 360° ANSI cut resistance.
  • ANSI cut level A5, abrasion resistance level 3, and puncture resistance level 3
  • Padded palm dampens impact for better control
  • Sandy nitrile palm coating for secure grip
  • Form-fitting shell delivers comfort and cut protection
  • High-visibility color enhances worker safety
  • Color-coded overcast reduces fraying and aids in sizing
  • Latex free
Catalog No. 50-006-88894 Supplier MSC Supplier No. 20867800
These size 7, 15-gauge seamless machine knit gloves feature a high-visibility yellow DuraBlend™ shell for breathability and comfort. The sandy NitriX® Grip palm coating ensures a secure grip in wet, dry, or oily applications, while the padded palm provides impact dampening for improved control. Designed for use in assembly, manufacturing, and material handling, these gloves provide 360° ANSI cut resistance.
  • ANSI cut level A5, abrasion resistance level 3, and puncture resistance level 3
  • Padded palm dampens impact for better control
  • Sandy nitrile palm coating for secure grip
  • Form-fitting shell delivers comfort and cut protection
  • High-visibility color enhances worker safety
  • Color-coded overcast reduces fraying and aids in sizing
  • Latex free