Learn More
MSC MAXIKUT 5 CR KNIT/NYL GLV
Catalog No. 502737423
Encompass
Cut-Resistant Gloves: Size M, ANSI Cut A4, Nitrile, Dyneema
Catalog No. 50-273-7423 Supplier MSC Supplier No. 70645577
Cut-Resistant Gloves: Size M, ANSI Cut A4, Nitrile, Dyneema