Learn More
Cayman Chemical Nitro Blue Tetrazolium (chloride), CAS: 298-83-9, 500 mg
SDP
Catalog No. 502966877
Encompass_Preferred
A chromogenic substrate that, like other tetrazolium compounds, can be reduced to produce a colored formazan derivative; used in the “NBT test” to evaluate the activity of NADPH oxidase in phagocytes; paired with PCIB for the colorimetric detection of alkaline phosphatase activity
Catalog No. 50-296-6877 Supplier Cayman Chemical Supplier No. 17341500 MG
A chromogenic substrate that, like other tetrazolium compounds, can be reduced to produce a colored formazan derivative; used in the “NBT test” to evaluate the activity of NADPH oxidase in phagocytes; paired with PCIB for the colorimetric detection of alkaline phosphatase activity